Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Elovl2
Entrez Gene ID
54326
Gene Name
elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 2
Gene Aliases
AI317360, Ssc2
Chromosome Location
Chr.13: 41182382 - 41220403 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019423.2 NP_062296.1 7 - 8 813 67
GenBank mRNA AF170908.1 - 7 - 8 803 67
AK014803.1 - 7 - 8 892 67
AK029855.1 - 7 - 8 794 67
AK044770.1 - 7 - 8 816 67
AK050397.1 - 7 - 8 816 67
BC098215.1 - 7 - 8 796 67
KF987080.1 - 5 - 6 711 67

View Full Product Details