Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jarid2
Entrez Gene ID
16468
Gene Name
jumonji, AT rich interactive domain 2
Gene Aliases
Jmj, jumonji
Chromosome Location
Chr.13: 44729181 - 44921643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205043.1 NP_001191972.1 1 - 2 646 137
NM_001205044.1 NP_001191973.1 2 - 3 319 137
NM_021878.3 NP_068678.1 2 - 3 365 137
GenBank mRNA AK044888.1 - 1 - 2 550 137
AK045214.1 - 1 - 2 451 137
AK147226.1 - 1 - 2 646 136
BC026056.1 - 1 - 2 553 137
BC050129.1 - 1 - 2 567 137
BC051010.1 - 1 - 2 549 137
BC052444.1 - 2 - 3 236 137
BC060695.1 - 2 - 3 236 137
D31967.1 - 2 - 3 255 137

View Full Product Details