Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jarid2
Entrez Gene ID
16468
Gene Name
jumonji, AT rich interactive domain 2
Gene Aliases
Jmj, jumonji
Chromosome Location
Chr.13: 44729181 - 44921643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205043.1 NP_001191972.1 7 - 8 2547 88
NM_001205044.1 NP_001191973.1 8 - 9 2220 88
NM_021878.3 NP_068678.1 8 - 9 2266 88
GenBank mRNA AK045214.1 - 7 - 8 2352 88
AK147226.1 - 7 - 8 2547 88
AK164134.1 - 6 - 7 1871 88
BC003374.1 - 1 - 2 116 88
BC026056.1 - 7 - 8 2454 88
BC050129.1 - 7 - 8 2468 88
BC051010.1 - 7 - 8 2450 88
BC052444.1 - 8 - 9 2137 88
BC060695.1 - 8 - 9 2137 88
BC115758.1 - 2 - 3 1151 88
D31967.1 - 8 - 9 2156 88

View Full Product Details