Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Akr1c19
- Entrez Gene ID
-
432720
- Gene Name
- aldo-keto reductase family 1, member C19
- Gene Aliases
-
1810010N06Rik
- Chromosome Location
-
Chr.13: 4233504 - 4248361 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BG086571.2 |
- |
- |
106 |
81 |
View Full Product Details