Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc85c
Entrez Gene ID
668158
Gene Name
coiled-coil domain containing 85C
Gene Aliases
EG668158, Gm265, Gm9010
Chromosome Location
Chr.12: 108203599 - 108275671 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159910.1 NP_001153382.1 1 - 1 690 76

View Full Product Details