The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007576.2 | NP_001007577.1 | 15 - 16 | 3198 | 58 |
GenBank mRNA | AY651761.1 | - | 15 - 16 | 3193 | 58 |
BC115714.1 | - | 14 - 15 | 3028 | 58 |