Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gucy2f
Entrez Gene ID
245650
Gene Name
guanylate cyclase 2f
Gene Aliases
A930002I06, GC-F, GC2
Chromosome Location
Chr.X: 142079286 - 142197276 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007576.2 NP_001007577.1 16 - 17 3299 67
GenBank mRNA AY651761.1 - 16 - 17 3294 67
BC115714.1 - 15 - 16 3129 67

View Full Product Details