Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx10
Entrez Gene ID
71982
Gene Name
sorting nexin 10
Gene Aliases
2410004M09Rik
Chromosome Location
Chr.6: 51523902 - 51590679 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127348.1 NP_001120820.1 2 - 3 175 89
NM_001127349.1 NP_001120821.1 2 - 3 180 89
NM_028035.4 NP_082311.3 2 - 3 278 89
GenBank mRNA AK010399.1 - 2 - 3 123 89
AK028163.1 - 2 - 3 279 89
AK150704.1 - 2 - 3 183 89
AK150878.1 - 2 - 3 97 89
AK150999.1 - 2 - 3 183 89
AK151320.1 - 2 - 3 183 89
AK151520.1 - 2 - 3 183 89
AK152004.1 - 2 - 3 101 89
AK152825.1 - 2 - 3 155 89
AK152949.1 - 2 - 3 171 89
AK153127.1 - 2 - 3 183 89
AK153356.1 - 2 - 3 133 89
AK166121.1 - 2 - 3 148 89
AK169110.1 - 2 - 3 114 89
BC010334.1 - 2 - 3 112 89

View Full Product Details