The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204427.1 | NP_001191356.1 | 2 - 2 | 710 | 96 |
NM_011478.2 | NP_035608.1 | 2 - 2 | 846 | 96 | |
GenBank mRNA | BC125629.1 | - | 1 - 1 | 740 | 96 |