Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mpp6
Entrez Gene ID
56524
Gene Name
membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6)
Gene Aliases
P55t, Pals2
Chromosome Location
Chr.6: 50110241 - 50198598 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164734.1 NP_001158206.1 1 - 2 224 115
NM_019939.2 NP_064323.2 1 - 2 224 115
GenBank mRNA AF199009.1 - 1 - 2 220 115
AF199010.1 - 1 - 2 220 115
AK144434.1 - 1 - 2 225 115
BC042868.2 - 1 - 2 171 115
BC046421.1 - 1 - 2 171 115
BC069908.1 - 1 - 2 213 115
BC106140.1 - 1 - 2 171 115
BC138666.1 - 1 - 2 82 115
BC145363.1 - 1 - 2 82 115

View Full Product Details