Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dfna5
- Entrez Gene ID
-
54722
- Gene Name
- deafness, autosomal dominant 5 (human)
- Gene Aliases
-
2310037D07Rik, 4932441K13Rik, Dfna5h, EG14210, Fin15
- Chromosome Location
-
Chr.6: 50207403 - 50262862 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016561.1 |
- |
- |
219 |
138 |
View Full Product Details