The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290986.1 | NP_001277915.1 | 11 - 12 | 1615 | 71 |
NM_001290987.1 | NP_001277916.1 | 10 - 11 | 1500 | 71 | |
NM_030234.2 | NP_084510.2 | 11 - 12 | 1630 | 71 | |
GenBank mRNA | AK031592.1 | - | 11 - 12 | 1556 | 71 |
AK162181.1 | - | 11 - 12 | 1583 | 71 | |
BC094676.1 | - | 10 - 11 | 1266 | 71 | |
BC117821.1 | - | 10 - 11 | 1439 | 71 | |
BC117822.1 | - | 11 - 12 | 1569 | 71 |