Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Duox2
Entrez Gene ID
214593
Gene Name
dual oxidase 2
Gene Aliases
A430065P05Rik, LNOX2, NOXEF2, P138-TOX, THOX2
Chromosome Location
Chr.2: 122279246 - 122298742 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177610.2 NP_808278.2 21 - 22 2914 81

View Full Product Details