Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tax1bp1
Entrez Gene ID
52440
Gene Name
Tax1 (human T cell leukemia virus type I) binding protein 1
Gene Aliases
1200003J11Rik, 1700069J21Rik, AA930106, D6Ertd404e, D6Ertd772e, T6bp, TXBP151
Chromosome Location
Chr.6: 52713575 - 52766780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025816.3 NP_080092.2 13 - 14 1905 149
GenBank mRNA AK004574.1 - 13 - 14 1887 149
AK146076.1 - 13 - 14 1885 149
BC014798.1 - 13 - 14 1826 149

View Full Product Details