Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sel1l
Entrez Gene ID
20338
Gene Name
sel-1 suppressor of lin-12-like (C. elegans)
Gene Aliases
AW493766, Sel1h, mKIAA4137
Chromosome Location
Chr.12: 91806043 - 91849157 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039089.1 NP_001034178.1 18 - 19 1988 83
NM_011344.2 NP_035474.1 18 - 19 1838 83
GenBank mRNA AF063095.1 - 18 - 19 1787 83
AK005023.2 - 18 - 19 1961 83
AK160936.1 - 18 - 19 1796 83
AK220502.1 - 18 - 19 1988 83
BC026816.1 - 18 - 19 1810 83
BC049959.1 - 13 - 14 1200 83
BC057452.1 - 4 - 5 423 83

View Full Product Details