The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288618.1 | NP_001275547.1 | 12 - 13 | 1330 | 63 |
NM_001288619.1 | NP_001275548.1 | 11 - 12 | 1237 | 63 | |
NM_001288620.1 | NP_001275549.1 | 10 - 11 | 1127 | 63 | |
NM_009953.4 | NP_034083.2 | 12 - 13 | 1327 | 63 | |
GenBank mRNA | AK142261.1 | - | 12 - 13 | 1277 | 63 |
AY445512.1 | - | 11 - 12 | 1237 | 63 | |
AY753668.1 | - | 10 - 11 | 1199 | 63 | |
BC137592.1 | - | 12 - 13 | 1213 | 63 | |
U17858.1 | - | 12 - 13 | 1155 | 63 |