The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199143.2 | NP_954594.1 | 1 - 2 | 559 | 143 |
GenBank mRNA | AF513708.1 | - | 1 - 2 | 559 | 143 |
BC104316.2 | - | 1 - 2 | 204 | 143 | |
BC104317.2 | - | 1 - 2 | 213 | 143 | |
BC125242.1 | - | 1 - 2 | 213 | 143 |