Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpvl
Entrez Gene ID
71287
Gene Name
carboxypeptidase, vitellogenic-like
Gene Aliases
4933436L16Rik, HVLP
Chromosome Location
Chr.6: 53873279 - 53978673 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289714.1 NP_001276643.1 8 - 9 793 102
GenBank mRNA AK017087.1 - 8 - 9 794 102

View Full Product Details