The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029981.1 | NP_084257.1 | 18 - 19 | 2949 | 69 |
GenBank mRNA | AB096928.1 | - | 18 - 19 | 2949 | 69 |
AK020831.1 | - | 2 - 3 | 411 | 69 | |
AK156991.1 | - | 18 - 19 | 2952 | 69 | |
AK172978.1 | - | 6 - 7 | 5355 | 69 | |
AK181027.1 | - | - | 119 | 69 | |
BC128568.1 | - | 18 - 19 | 2901 | 69 | |
BC129984.1 | - | 18 - 19 | 2901 | 69 |