Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adamtsl2
Entrez Gene ID
77794
Gene Name
ADAMTS-like 2
Gene Aliases
A930008K15Rik, tcp-1
Chromosome Location
Chr.2: 27079342 - 27108613 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029981.1 NP_084257.1 2 - 3 287 81
GenBank mRNA AB096928.1 - 2 - 3 287 81
AK156991.1 - 2 - 3 290 81
BC128568.1 - 2 - 3 239 81
BC129984.1 - 2 - 3 239 81

View Full Product Details