Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a6
Entrez Gene ID
227659
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 6
Gene Aliases
A330096C23, F630103L12Rik, Glut6, Glut9
Chromosome Location
Chr.2: 27021363 - 27028001 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177627.1 NP_001171098.1 1 - 2 179 60
NM_172659.2 NP_766247.2 1 - 2 179 60
GenBank mRNA AK079650.1 - 1 - 2 123 60
AK089246.1 - 1 - 2 171 60
AK137441.1 - 1 - 2 145 60
AK150104.1 - 1 - 2 132 60
AK163445.1 - 1 - 2 141 60
AK166312.1 - 1 - 2 137 60
AK167904.1 - 1 - 2 141 60
AK172411.1 - 1 - 2 123 60
BC058210.1 - 1 - 2 103 60
BC141168.1 - 1 - 2 110 60
BC145480.1 - 1 - 2 110 60
BY202464.1 - 1 - 2 180 60

View Full Product Details