Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a6
Entrez Gene ID
227659
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 6
Gene Aliases
A330096C23, F630103L12Rik, Glut6, Glut9
Chromosome Location
Chr.2: 27021363 - 27028001 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177627.1 NP_001171098.1 5 - 6 860 87
NM_172659.2 NP_766247.2 5 - 6 860 87
GenBank mRNA AK079650.1 - 5 - 6 804 87
AK089246.1 - 5 - 6 852 87
AK150104.1 - 5 - 6 813 87
AK166312.1 - 5 - 6 818 87
AK167904.1 - 5 - 6 822 87
BC058210.1 - 5 - 6 784 87
BC141168.1 - 5 - 6 791 87

View Full Product Details