Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rif1
Entrez Gene ID
51869
Gene Name
Rap1 interacting factor 1 homolog (yeast)
Gene Aliases
5730435J01Rik, 6530403D07Rik, AU016181, AW549474, D2Ertd145e
Chromosome Location
Chr.2: 52072811 - 52126769 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175238.5 NP_780447.4 19 - 20 2153 124
GenBank mRNA AK088688.1 - 7 - 8 1845 124
AY428571.2 - 19 - 20 2137 124
AY585206.1 - 18 - 19 2146 124
JX255737.1 - 18 - 19 2089 124

View Full Product Details