The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028923.3 | NP_083199.1 | 1 - 2 | 187 | 75 |
GenBank mRNA | AK016671.1 | - | 1 - 2 | 169 | 75 |
AK028183.1 | - | 1 - 2 | 171 | 75 | |
AK029395.1 | - | 1 - 2 | 141 | 75 | |
AK133888.1 | - | 1 - 2 | 139 | 75 | |
AK145748.1 | - | 1 - 2 | 163 | 75 | |
AK150230.1 | - | 1 - 2 | 162 | 75 | |
AK161015.1 | - | 1 - 2 | 133 | 75 |