Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trub2
Entrez Gene ID
227682
Gene Name
TruB pseudouridine (psi) synthase homolog 2 (E. coli)
Gene Aliases
G430055L02Rik
Chromosome Location
Chr.2: 29761223 - 29787810 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290495.1 NP_001277424.1 5 - 6 528 71
NM_145520.5 NP_663495.3 6 - 7 590 71
GenBank mRNA AK043949.1 - 6 - 7 585 71
AK050587.1 - 5 - 6 518 71
AK081579.1 - 5 - 6 518 71
AK090004.1 - - 1422 71
AK167294.1 - 6 - 7 566 71
BC015285.1 - 6 - 7 569 71
BC024075.1 - 6 - 7 559 71

View Full Product Details