Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdia4
Entrez Gene ID
12304
Gene Name
protein disulfide isomerase associated 4
Gene Aliases
AI987846, Cai, ERp-72, Erp72
Chromosome Location
Chr.6: 47796141 - 47813512 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009787.2 NP_033917.2 8 - 9 1487 70
GenBank mRNA AK028292.1 - 8 - 9 1337 70
AK146288.1 - 8 - 9 1320 70
AK148801.1 - 8 - 9 1409 70
AK150566.1 - 8 - 9 1368 70
AK161534.1 - 8 - 9 1325 70
AK168447.1 - 8 - 9 1291 70
AK169387.1 - 8 - 9 1332 70
BC066857.1 - 7 - 8 1086 70
BC141078.1 - 8 - 9 1383 70
J05186.1 - 8 - 9 1370 70
Y00884.1 - 7 - 8 1193 70

View Full Product Details