The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175048.3 | NP_778213.2 | 4 - 5 | 313 | 131 |
GenBank mRNA | AK088228.1 | - | 4 - 5 | 431 | 131 |