The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207298.2 | NP_997181.1 | 3 - 4 | 465 | 95 |
GenBank mRNA | AK155384.1 | - | 3 - 4 | 461 | 95 |
BC052176.1 | - | 3 - 4 | 426 | 95 | |
BC138847.1 | - | 3 - 4 | 444 | 95 |