Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptprj
Entrez Gene ID
19271
Gene Name
protein tyrosine phosphatase, receptor type, J
Gene Aliases
AI450271, BET, Byp, CD148, DEP-1, PTPbeta2, Ptpb2, RPTPJ, Scc-1, Scc1
Chromosome Location
Chr.2: 90429756 - 90580677 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135657.1 NP_001129129.1 22 - 23 3535 81
NM_008982.5 NP_033008.3 22 - 23 3819 81
GenBank mRNA AK131723.1 - 4 - 5 475 81
AK147318.1 - 22 - 23 3740 81
AK147556.1 - 22 - 23 3740 81
AY038891.1 - 22 - 23 3637 81
AY039232.1 - 22 - 23 3637 81
D45212.1 - 22 - 23 3775 81
D49393.1 - 9 - 10 792 81
D83203.1 - 22 - 23 3536 81
DQ133576.1 - 22 - 23 3550 81

View Full Product Details