The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177602.3 | NP_808270.2 | 3 - 4 | 1007 | 91 |
GenBank mRNA | AK147993.1 | - | 4 - 5 | 2709 | 91 |