Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp8b4
Entrez Gene ID
241633
Gene Name
ATPase, class I, type 8B, member 4
Gene Aliases
A530043E15, Im
Chromosome Location
Chr.2: 126320973 - 126500669 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080944.3 NP_001074413.2 18 - 19 2037 161
GenBank mRNA AK086607.1 - 17 - 18 162
AK155448.1 - 18 - 19 2047 161
AK169967.1 - 18 - 19 2060 166
AK173307.1 - 18 - 19 2037 161

View Full Product Details