The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024616.1 | NP_001019787.2 | 12 - 13 | 1961 | 105 |
GenBank mRNA | AK045520.1 | - | 12 - 13 | 1983 | 105 |
AK081440.1 | - | 12 - 13 | 1971 | 105 | |
AK157020.1 | - | 12 - 13 | 1996 | 105 | |
AK162461.1 | - | 8 - 9 | 1341 | 105 | |
AK171643.1 | - | 12 - 13 | 1972 | 105 | |
BC006855.1 | - | 4 - 5 | 3693 | 105 | |
BC020111.1 | - | 12 - 13 | 1969 | 105 | |
BC021476.1 | - | 12 - 13 | 1976 | 105 | |
BC023692.1 | - | 12 - 13 | 1989 | 105 | |
BC040791.1 | - | 4 - 5 | 3693 | 105 | |
BC051138.1 | - | 12 - 13 | 1975 | 105 | |
BC058585.1 | - | 12 - 13 | 1974 | 105 | |
BC085507.1 | - | 12 - 13 | 1961 | 105 |