The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199141.1 | NP_001186070.1 | 7 - 8 | 972 | 92 |
NM_144516.3 | NP_653099.2 | 6 - 7 | 810 | 92 | |
GenBank mRNA | AK028931.1 | - | 4 - 5 | 394 | 92 |
AK077732.1 | - | 6 - 7 | 657 | 92 | |
AK145892.1 | - | 6 - 7 | 604 | 92 | |
BC022945.1 | - | 7 - 8 | 789 | 92 | |
CT010329.1 | - | 5 - 6 | 573 | 92 |