The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252454.1 | NP_001239383.1 | 10 - 11 | 1790 | 71 |
NM_010147.4 | NP_034277.1 | 11 - 12 | 1961 | 71 | |
GenBank mRNA | AF057285.1 | - | 3 - 4 | 434 | 71 |
AK155033.1 | - | 11 - 12 | 1957 | 71 | |
BC046962.1 | - | 9 - 10 | 1596 | 71 | |
BC067206.1 | - | 10 - 11 | 1732 | 71 | |
BC099682.1 | - | 11 - 12 | 1891 | 71 |