Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ap4e1
Entrez Gene ID
108011
Gene Name
adaptor-related protein complex AP-4, epsilon 1
Gene Aliases
2310033A20Rik, 9930028M04Rik, AV087807
Chromosome Location
Chr.2: 127008679 - 127069814 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175550.3 NP_780759.2 15 - 16 1998 87
GenBank mRNA AK036944.1 - 15 - 16 1979 87
AK144636.1 - 15 - 16 1979 87
BC042530.1 - 2 - 3 189 87

View Full Product Details