Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mastl
Entrez Gene ID
67121
Gene Name
microtubule associated serine/threonine kinase-like
Gene Aliases
2700091H24Rik, C88295, GW, GWL, MAST-L, THC2
Chromosome Location
Chr.2: 23116544 - 23156024 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025979.4 NP_080255.3 7 - 8 1140 77
GenBank mRNA AK006448.1 - 2 - 3 241 77
AK012597.1 - 2 - 3 241 77
AK030140.1 - 7 - 8 1035 77
BC066834.1 - - 397 77
BC086483.1 - - 409 77
BC103779.1 - 7 - 8 1016 77
CD546688.1 - 3 - 4 363 77

View Full Product Details