The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077190.2 | NP_001070658.1 | 9 - 10 | 1266 | 107 |
NM_007380.3 | NP_031406.2 | 8 - 9 | 1248 | 107 | |
NM_145994.2 | NP_666106.1 | 8 - 9 | 1251 | 107 | |
GenBank mRNA | AF420251.1 | - | 8 - 9 | 1184 | 107 |
AK034476.1 | - | 9 - 10 | 1148 | 107 | |
AK167328.1 | - | 8 - 9 | 1207 | 107 | |
AK167568.1 | - | 8 - 9 | 1206 | 107 | |
AK179494.1 | - | 2 - 3 | 150 | 105 | |
AK182193.1 | - | 2 - 3 | 152 | 107 | |
AK186471.1 | - | 2 - 3 | 153 | 107 | |
BC004657.1 | - | 8 - 9 | 1202 | 107 |