The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019456.2 | NP_062329.2 | 13 - 14 | 1689 | 86 |
GenBank mRNA | AF020313.1 | - | 10 - 11 | 1293 | 86 |
AK089987.1 | - | 5 - 6 | 563 | 86 | |
BC003991.1 | - | 10 - 11 | 1340 | 86 | |
BC023110.1 | - | 13 - 14 | 1578 | 86 |