Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hspa12b
Entrez Gene ID
72630
Gene Name
heat shock protein 12B
Gene Aliases
2700081N06Rik
Chromosome Location
Chr.2: 131127412 - 131145985 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028306.3 NP_082582.1 10 - 11 1161 57
GenBank mRNA AK012548.1 - 10 - 11 1134 57
AY196790.1 - 9 - 10 1041 57
BC011103.1 - 10 - 11 1161 57

View Full Product Details