Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc39a13
Entrez Gene ID
68427
Gene Name
solute carrier family 39 (metal ion transporter), member 13
Gene Aliases
1100001L14Rik, AA387663, ZIP-13, ZIP13
Chromosome Location
Chr.2: 91061781 - 91070315 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290765.1 NP_001277694.1 4 - 5 744 97
NM_026721.3 NP_080997.1 4 - 5 705 97
GenBank mRNA AK035296.1 - 4 - 5 612 97
AK156951.1 - 4 - 5 630 97
BC100463.1 - 4 - 5 712 97
BC125525.1 - 4 - 5 566 97
BC137752.1 - 4 - 5 566 97

View Full Product Details