Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdia3
Entrez Gene ID
14827
Gene Name
protein disulfide isomerase associated 3
Gene Aliases
58kDa, ERp57, ERp60, ERp61, Erp, Grp58, PDI, PDI-Q2, PI-PLC, PLC[a], Plca
Chromosome Location
Chr.2: 121413902 - 121438687 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007952.2 NP_031978.2 5 - 6 733 125
GenBank mRNA AK088721.1 - 5 - 6 693 125
AK132459.1 - 5 - 6 698 125
AK133799.1 - 5 - 6 692 125
AK167807.1 - 5 - 6 686 125
AK169611.1 - 5 - 6 687 125
BC003285.1 - 5 - 6 652 125
BC033439.1 - 5 - 6 627 125

View Full Product Details