Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prpf31
Entrez Gene ID
68988
Gene Name
PRP31 pre-mRNA processing factor 31 homolog (yeast)
Gene Aliases
1500019O16Rik, 2810404O06Rik, AW554706, PRP31, RP11
Chromosome Location
Chr.7: 3629985 - 3642485 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159714.1 NP_001153186.1 4 - 5 399 103
NM_027328.4 NP_081604.3 4 - 5 399 103
GenBank mRNA AK005294.1 - 4 - 5 367 103
AK033283.1 - 4 - 5 364 103
AK044457.1 - 4 - 5 462 103
AK051260.1 - 4 - 5 359 103
AY040823.1 - 4 - 5 375 103
BC018376.1 - 4 - 5 365 103
BC057877.1 - 3 - 4 342 103
BC061461.1 - 4 - 5 343 103
CT010189.1 - 3 - 4 319 103

View Full Product Details