Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thnsl1
Entrez Gene ID
208967
Gene Name
threonine synthase-like 1 (bacterial)
Gene Aliases
AW413632
Chromosome Location
Chr.2: 21205385 - 21215009 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001297.2 NP_001001297.1 1 - 2 122 84
NM_177588.2 NP_808256.1 1 - 2 122 84
GenBank mRNA AK029518.1 - 1 - 2 123 84
AK033465.1 - 1 - 2 91 84
AK083895.1 - 1 - 2 122 84

View Full Product Details