Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lcn11
Entrez Gene ID
227630
Gene Name
lipocalin 11
Gene Aliases
Gm109
Chromosome Location
Chr.2: 25777017 - 25780279 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100455.2 NP_001093925.1 5 - 6 534 76
GenBank mRNA BE864901.1 - 4 - 5 352 76

View Full Product Details