Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lhx3
Entrez Gene ID
16871
Gene Name
LIM homeobox protein 3
Gene Aliases
Lim3, P-LIM, mLIM3, mLim-3
Chromosome Location
Chr.2: 26200212 - 26208249 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039653.2 NP_001034742.1 4 - 5 754 58
NM_010711.2 NP_034841.2 4 - 5 671 58
GenBank mRNA AK047273.1 - 1 - 2 514 58
BC132554.1 - 4 - 5 668 58
BC132556.1 - 4 - 5 668 58
BC150689.1 - 4 - 5 640 58
L38248.1 - 4 - 5 720 58
L38857.1 - 4 - 5 671 58

View Full Product Details