The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205231.1 | NP_001192160.1 | 5 - 6 | 627 | 59 |
NM_133671.3 | NP_598432.2 | 5 - 6 | 627 | 59 | |
GenBank mRNA | AK078496.1 | - | 5 - 6 | 581 | 59 |
BC043071.1 | - | 5 - 6 | 627 | 59 | |
BC094451.1 | - | 3 - 4 | 280 | 59 | |
BU054258.1 | - | 5 - 6 | 627 | 59 | |
X64587.1 | - | 5 - 6 | 533 | 59 |