The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201355.3 | NP_958743.1 | 2 - 3 | 506 | 82 |
GenBank mRNA | AK078272.1 | - | 2 - 3 | 172 | 82 |
AK158627.1 | - | 2 - 3 | 507 | 82 | |
AK163549.1 | - | 2 - 3 | 234 | 82 | |
BC047219.1 | - | 2 - 3 | 142 | 82 |