Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnnt1
Entrez Gene ID
21955
Gene Name
troponin T1, skeletal, slow
Gene Aliases
AW146156, Tnt, sTnT, ssTnT
Chromosome Location
Chr.7: 4504570 - 4515999 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277903.1 NP_001264832.1 1 - 2 48 69
NM_001277904.1 NP_001264833.1 1 - 2 48 69
NM_011618.2 NP_035748.1 1 - 2 48 69
GenBank mRNA AK160226.1 - 1 - 2 49 69

View Full Product Details