The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139200.4 | NP_631939.1 | 6 - 7 | 614 | 72 |
GenBank mRNA | AF192525.1 | - | 6 - 7 | 534 | 72 |
AK152784.1 | - | 6 - 7 | 596 | 72 | |
AK157859.1 | - | 6 - 7 | 615 | 72 | |
AK166254.1 | - | 6 - 7 | 615 | 72 | |
AK169575.1 | - | 6 - 7 | 600 | 72 | |
AK170544.1 | - | 6 - 7 | 615 | 72 | |
AK170796.1 | - | 6 - 7 | 615 | 72 | |
AK171467.1 | - | 6 - 7 | 615 | 72 | |
AK172071.1 | - | 6 - 7 | 671 | 72 | |
BC007144.1 | - | 6 - 7 | 603 | 72 |