Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Brf1
Entrez Gene ID
72308
Gene Name
BRF1 homolog, subunit of RNA polymerase III transcription initiation factor IIIB (S. cerevisiae)
Gene Aliases
2510002F24Rik, GTF3B, TAF3C, TAFIII90, TFIIIB90, mTFIIIB90
Chromosome Location
Chr.12: 112955631 - 113000707 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028193.3 NP_082469.2 16 - 17 2081 60
GenBank mRNA AK010890.1 - 3 - 4 330 60
BC037224.1 - 16 - 17 1864 60
BC058112.1 - 6 - 7 677 60

View Full Product Details